missing translation for 'onlineSavingsMsg'
Learn More

2,2-Diphenylpropylamine hydrochloride, 98+%

Product Code. 11328356
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11328356 1 g 1g
11338356 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11328356 Supplier Thermo Scientific Alfa Aesar Supplier No. L15615.03
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 40691-66-5 | C15H18ClN | 247.766 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 40691-66-5
Molecular Formula C15H18ClN
Molecular Weight (g/mol) 247.766
MDL Number MFCD00008133
InChI Key AASCJSPDUDWGGQ-UHFFFAOYSA-N
Synonym 2,2-diphenylpropylamine hydrochloride, 2,2-diphenylpropan-1-amine hydrochloride, beta-methyl-beta-phenylphenethylamine hydrochloride, acmc-20aojd, diphenylpropylamine hci, beta-methyl-beta-phenylphenethylamine hcl
PubChem CID 3084798
IUPAC Name 2,2-diphenylpropan-1-amine;hydrochloride
SMILES CC(CN)(C1=CC=CC=C1)C2=CC=CC=C2.Cl

Specifications

Melting Point 250°C to 256°C
Quantity 1 g
Beilstein 3709987
Formula Weight 247.77
Percent Purity ≥98%
Chemical Name or Material 2,2-Diphenylpropylamine hydrochloride

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.