missing translation for 'onlineSavingsMsg'
Learn More

2,2-Diphenylglycine, 98%

Product Code. 11445267
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11445267 100 g 100g
11425267 5 g 5g
11435267 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11445267 Supplier Thermo Scientific Alfa Aesar Supplier No. B22650.22
 Restricted Item
Estimated Shipment: 04-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 3060-50-2 | C14H13NO2 | 227.26 g/mol

2,2-Diphenylglycine is used as a synthesis reagent for amination reactions. It acts as an intermediate in pharmaceutical and in organic synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,2-Diphenylglycine is used as a synthesis reagent for amination reactions. It acts as an intermediate in pharmaceutical and in organic synthesis.

Solubility
It is sparingly soluble in water.

Notes
Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3060-50-2
Molecular Formula C14H13NO2
Molecular Weight (g/mol) 227.26
MDL Number MFCD00008048
InChI Key YBONNYNNFBAKLI-UHFFFAOYSA-N
Synonym 2,2-diphenylglycine, alpha,alpha-diphenylglycine, alpha-aminodiphenylacetic acid, glycine, 2,2-diphenyl, alpha-amino-alpha-phenylbenzeneacetic acid, .alpha.-aminodiphenylacetic acid, .alpha.,.alpha.-diphenylglycine, benzeneacetic acid, .alpha.-amino-.alpha.-phenyl, aminodiphenylacetic acid, phenytoin impurity c
PubChem CID 18289
IUPAC Name 2-amino-2,2-diphenylacetic acid
SMILES NC(C(O)=O)(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point ∼246°C (decomposition)
Solubility Information It is sparingly soluble in water.
Quantity 100 g
Formula Weight 227.26
Percent Purity 98%
Chemical Name or Material 2,2-Diphenylglycine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.