missing translation for 'onlineSavingsMsg'
Learn More

2,2-Diphenylethanol, 97%

Product Code. 10274784 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
2 g
10 g
Packaging:
Glass bottle
Unit Size:
10g
2g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10274784 2 g Glass bottle 2g
10153354 10 g Glass bottle 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10274784 Supplier Thermo Scientific Acros Supplier No. 188220020
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 1883-32-5 | C14H14O | 198.26 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1883-32-5
Molecular Formula C14H14O
Molecular Weight (g/mol) 198.26
MDL Number MFCD00004729
InChI Key NYLOEXLAXYHOHH-UHFFFAOYSA-N
Synonym beta-phenylphenethyl alcohol, 2,2-diphenyl ethanol, benzeneethanol, .beta.-phenyl, acmc-20aojg, 2,2-diphenyl-ethanol, 2,2-diphenyl-1-ethanol, 2,2-diphenylethan-1-ol, benzeneethanol, b-phenyl, 2,2-diphenylethanol, ksc539o6l
PubChem CID 74662
IUPAC Name 2,2-diphenylethanol
SMILES C1=CC=C(C=C1)C(CO)C2=CC=CC=C2

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 53°C to 56°C
Color White
Boiling Point 160°C to 162°C (5.0 mmHg)
Flash Point >110°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Linear Formula (C6H5)2CHCH2OH
Quantity 2 g
Beilstein 06, II, 640
Formula Weight 198.26
Percent Purity 97%
Physical Form Crystalline Powder
Chemical Name or Material 2, 2-Diphenylethanol
Mostrar más Mostrar menos
compliance-icons
Nombre del producto
  • 2,2-Diphenylethanol
Palabra de advertencia
  • Warning
Clasificación
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Indicaciones de peligro
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Consejos de prudencia
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.