missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,2-Diphenylcyclopropanecarboxylic Acid, TRC
Specifications
Specifications
| Chemical Name or Material | 2,2-Diphenylcyclopropanecarboxylic Acid |
| CAS | 7150-12-1 |
| Molecular Formula | C16 H14 O2 |
| Quantity | 250 mg |
| Synonym | NSC 120410, NSC 74083 |
| InChI Formula | InChI=1S/C16H14O2/c17-15(18)14-11-16(14,12-7-3-1-4-8-12)13-9-5-2-6-10-13/h1-10,14H,11H2,(H,17,18) |
| SMILES | OC(=O)C1CC1(c2ccccc2)c3ccccc3 |
| IUPAC Name | 2,2-diphenylcyclopropane-1-carboxylic acid |
| Molecular Weight (g/mol) | 238.28 |
| Formula Weight | 238.0994 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?