missing translation for 'onlineSavingsMsg'
Learn More

2,2-Diphenylcyclopropanecarboxylic Acid, TRC

Product Code. 30373003
Change view
Click to view available options
Quantity:
100 mg
250 mg
Unit Size:
100mg
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30373003 100 mg 100mg
30385120 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30373003 Supplier Toronto Research Chemicals Supplier No. TRCD487515100MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 2,2-Diphenylcyclopropanecarboxylic Acid
CAS 7150-12-1
Molecular Formula C16 H14 O2
Quantity 100 mg
Synonym NSC 120410, NSC 74083
InChI Formula InChI=1S/C16H14O2/c17-15(18)14-11-16(14,12-7-3-1-4-8-12)13-9-5-2-6-10-13/h1-10,14H,11H2,(H,17,18)
SMILES OC(=O)C1CC1(c2ccccc2)c3ccccc3
IUPAC Name 2,2-diphenylcyclopropane-1-carboxylic acid
Molecular Weight (g/mol) 238.28
Formula Weight 238.0994
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.