Learn More
2,2-Diphenyl-1-picrylhydrazyl (free radical), 95%
Description
2,2-Diphenyl-1-picrylhydrazyl is used as a sensitive colorimetric free radical scavenger. It serves as a general antioxidant detector. It is utilized to monitor chemical reactions involving radicals as well as a standard of the position and intensity of electron paramagnetic resonance signals. Further, it is employed in the quantitative determination of aliphatic and aromatic thiols by indirect spectroscopy and for photometric determination of tocopherol.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Quantity | 1 g |
| Merck Index | 14,3330 |
| Solubility Information | Soluble in dimethyl formamide,ethanol,chloroform,ether,carbon disulfinde,acetone,oils,carbon tetrachloride,glacial acetic acid and toluene. Insoluble in water. |
| Formula Weight | 394.32 |
| Percent Purity | 95% |
| Physical Form | Powder |
| Chemical Name or Material | 2, 2-Diphenyl-1-picrylhydrazyl (Free Radical) |
Safety and Handling
- 2,2-Diphenyl-1-picrylhydrazyl (free radical)
- Danger
- Respiratory sensitiser Category 1
- Skin sensitiser Category 1
- H317-May cause an allergic skin reaction.
- H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P284-¢In case of inadequate ventilation! wear respiratory protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/soap.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P333+P313-If skin irritation or rash occurs: Get medical advice/attention.
- P342+P311-If experiencing respiratory symptoms: Call a POISON CENTER/doctor/ .
- MIXTURE LIST-Contain: 2,2-diphenyl-1-(2,4,6-trinitrophenyl)hydrazyl
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.