missing translation for 'onlineSavingsMsg'
Learn More

2,2'-Dinaphthyl ether, 98+%

Product Code. 11337745
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11337745 5 g 5g
11347745 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11337745 Supplier Thermo Scientific Alfa Aesar Supplier No. L02565.06
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 613-80-9 | C20H14O | 270.33 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 613-80-9
Molecular Formula C20H14O
Molecular Weight (g/mol) 270.33
MDL Number MFCD00015447
InChI Key DZRLNYVDCIYXPG-UHFFFAOYSA-N
PubChem CID 69183
IUPAC Name 2-naphthalen-2-yloxynaphthalene
SMILES O(C1=CC=C2C=CC=CC2=C1)C1=CC=C2C=CC=CC2=C1

Specifications

Melting Point 103°C to 105°C
Quantity 5 g
Beilstein 2054097
Synonym 2,2'-dinaphthyl ether, di-2-naphthyl ether, 2,2'-oxydinaphthalene, naphthalene, 2,2'-oxybis, bis 2-naphthyl ether, 2-naphthyl ether, di-2-naphthylether, 2,2'-dinaphthylether, naphthalene,2,2'-oxybis
Formula Weight 270.33
Percent Purity ≥98%
Chemical Name or Material 2,2'-Dinaphthyl ether

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.