missing translation for 'onlineSavingsMsg'
Learn More

2,2-Dimorpholinodiethylether, TRC

Product Code. 30385366
Change view
Click to view available options
Quantity:
250 mg
500 mg
2.5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30385366 250 mg
30384274 500 mg
30377955 2.5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30385366 Supplier Toronto Research Chemicals Supplier No. TRCD480378250MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6425-39-4
Molecular Formula C12H24N2O3
Molecular Weight (g/mol) 244.33
Synonym Morpholine, 4,4'-(oxydi-2,1-ethanediyl)bis- (9CI, ACI), 4,4'-(Oxydi-2,1-ethanediyl)bis[morpholine] (ACI), Morpholine, 4,4'-(oxydiethylene)di- (6CI, 7CI, 8CI), 2,2'-Dimorpholinodiethyl ether, 2,2'-Dimorpholinyldiethyl ether, 4,4'-(Oxydi-2,1-ethanediyl)bismorpholine, 4,4'-(Oxydiethylene)bis[morpholine], 4,4'-(Oxydiethylene)dimorpholine, B 802012, Bis(2-morpholinoethyl) ether, Bis(morpholinoethyl) ether, Bis[2-(4-morpholino)ethyl] ether, Dabco DMDEE, Di(2-morpholinoethyl) ether, Dimorpholinodiethyl ether, DMDEE, JD-DMDEE, Jeffcat DMDEE, Lupragen N 106, Maycat DMDEE, N 106, NSC 28749, PC CAT DMDEE, Texacat DMDEE, U-CAT 660M, β,β'-Dimorpholinodiethyl ether
IUPAC Name 4-[2-(2-morpholin-4-ylethoxy)ethyl]morpholine
SMILES C(CN1CCOCC1)OCCN2CCOCC2

Specifications

Quantity 250 mg
InChI Formula InChI=1S/C12H24N2O3/c1-7-15-8-2-13(1)5-11-17-12-6-14-3-9-16-10-4-14/h1-12H2
Formula Weight 244.1787
Chemical Name or Material 2,2-Dimorpholinodiethylether
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.