missing translation for 'onlineSavingsMsg'
Learn More

2,2-Dimethylcyclopentanone, 97%

Product Code. 10423274 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10423274 1 g Glass bottle 1g
10433274 5 g Glass bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10423274 Supplier Thermo Scientific Acros Supplier No. 399400010
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 4541-32-6 | C7H12O | 112.17 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4541-32-6
Molecular Formula C7H12O
Molecular Weight (g/mol) 112.17
MDL Number MFCD00049164
InChI Key FTGZMZBYOHMEPS-UHFFFAOYSA-N
Synonym 2,2-dimethylcyclopentanone, cyclopentanone, 2,2-dimethyl, cyclopentanone, dimethyl, zlchem 1228, 2,2 dimethylcyclopentanone, 2,2-dimethyl-cyclopentanone, 2,2-dimethylcyclo pentanone, 2,2-dimethylcyclo-pentanone, 2,2-dimethyl-cyclopentan-1-one
PubChem CID 138286
IUPAC Name 2,2-dimethylcyclopentan-1-one
SMILES CC1(CCCC1=O)C

Specifications

CAS Min % 96.0
CAS Max % 100.0
Color White to Yellow
Density 0.8900g/mL
Boiling Point 143°C to 145°C
Flash Point 32°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Linear Formula (CH3)2C5H6(=O)
Refractive Index 1.4320 to 1.4340 (20°C, 589nm)
Quantity 1 g
Specific Gravity 0.89
Formula Weight 112.17
Percent Purity 97%
Physical Form Solid
Chemical Name or Material 2, 2-Dimethylcyclopentanone
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 2,2-Dimethylcyclopentanone
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.