missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,2-Dimethylbutyryl chloride, 98%, Thermo Scientific™
Click to view available options
Quantity:
10 g
50 g
250 g
Unit Size:
10g
250g
50g
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Applications
,2-Dimethylbutanoyl chloride is a reagent that is used in the synthesis of 4α,5-Dihydro Simvastatin, the drug, is sold under the trade name Zocor. A competitive inhibitor of HMG-CoA reductase. A synthetic analog of Lovastatin. Antilipemic.
Solubility
Reacts with water. Soluble in chloroform and ethyl acetate.
Notes
Moisture sensitive. Store away from water/moisture, strong bases and oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Store under inert gas. Keep ignition sources away. Protect from electrostatic charges.
,2-Dimethylbutanoyl chloride is a reagent that is used in the synthesis of 4α,5-Dihydro Simvastatin, the drug, is sold under the trade name Zocor. A competitive inhibitor of HMG-CoA reductase. A synthetic analog of Lovastatin. Antilipemic.
Solubility
Reacts with water. Soluble in chloroform and ethyl acetate.
Notes
Moisture sensitive. Store away from water/moisture, strong bases and oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Store under inert gas. Keep ignition sources away. Protect from electrostatic charges.
Specifications
Specifications
| Density | 0.98 |
| Boiling Point | 133°C |
| Flash Point | 29°C (84°F) |
| Refractive Index | 1.424 |
| Quantity | 250 g |
| UN Number | UN2920 |
| Beilstein | 1743935 |
| Sensitivity | Moisture sensitive |
| Solubility Information | Reacts with water. Soluble in chloroform and ethyl acetate. |
| Formula Weight | 134.61 |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction