missing translation for 'onlineSavingsMsg'
Learn More

2,2-Dimethylbutyric acid, 99%

Product Code. 10724502 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Glass bottle
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10724502 25 g Glass bottle 25g
10139699 100 g Glass bottle 100g
10734502 500 g Glass bottle 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10724502 Supplier Thermo Scientific Acros Supplier No. 434860250
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 595-37-9 | C6H12O2 | 116.16 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 595-37-9
Molecular Formula C6H12O2
Molecular Weight (g/mol) 116.16
MDL Number MFCD00004200
InChI Key VUAXHMVRKOTJKP-UHFFFAOYSA-N
Synonym 2,2-dimethylbutyric acid, butanoic acid, 2,2-dimethyl, 2,2-dimethyl-butanoic acid, 2,2-dimethyl butyric acid, 2,2-dimethyl butanoic acid, unii-ay606cn05o, butyric acid, 2,2-dimethyl, dimethyl ethyl acetic acid, 2,2-dimethyl-butyric acid, .alpha.,.alpha.-dimethylbutyric acid
PubChem CID 11684
ChEBI CHEBI:38649
IUPAC Name 2,2-dimethylbutanoic acid
SMILES CCC(C)(C)C(=O)O

Specifications

CAS Min % 98.5
CAS Max % 100.0
Color Colorless to Yellow
Density 0.9280g/mL
Boiling Point 186.0°C to 187.0°C
Flash Point 79°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Quantity 25 g
Specific Gravity 0.928
Formula Weight 116.16
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material 2, 2-Dimethylbutyric acid
Show More Show Less
compliance-icons
Product Identifier
  • 2,2-Dimethylbutyric acid
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.