missing translation for 'onlineSavingsMsg'
Learn More

2,2-Dimethylbutyric acid, 97%

Product Code. 11428936
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
Unit Size:
10g
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11428936 10 g 10g
11438936 50 g 50g
11448936 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11428936 Supplier Thermo Scientific Alfa Aesar Supplier No. A17072.09
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 595-37-9 | C6H12O2 | 116.16 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 595-37-9
Molecular Formula C6H12O2
Molecular Weight (g/mol) 116.16
MDL Number MFCD00004200
InChI Key VUAXHMVRKOTJKP-UHFFFAOYSA-N
Synonym 2,2-dimethylbutyric acid, butanoic acid, 2,2-dimethyl, 2,2-dimethyl-butanoic acid, 2,2-dimethyl butyric acid, 2,2-dimethyl butanoic acid, unii-ay606cn05o, butyric acid, 2,2-dimethyl, dimethyl ethyl acetic acid, 2,2-dimethyl-butyric acid, .alpha.,.alpha.-dimethylbutyric acid
PubChem CID 11684
ChEBI CHEBI:38649
IUPAC Name 2,2-dimethylbutanoic acid
SMILES CCC(C)(C)C(=O)O

Specifications

Melting Point -14°C
Density 0.926
Boiling Point 186°C
Flash Point 90°C (194°F)
Odor Pungent
Refractive Index 1.4154
Quantity 10 g
UN Number UN3265
Beilstein 1720666
Formula Weight 116.16
Percent Purity 97%
Chemical Name or Material 2,2-Dimethylbutyric acid
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.