missing translation for 'onlineSavingsMsg'
Learn More

2-[2-(Dimethylamino)ethoxy]benzylamine, 90%, Thermo Scientific™

Product Code. 10205414
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10205414 1 g 1g
10525554 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10205414 Supplier Thermo Scientific Maybridge Supplier No. CC51313DA
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 91215-97-3
Molecular Formula C11H18N2O
Molecular Weight (g/mol) 194.28
MDL Number MFCD08690290
InChI Key NOZRJUDRDVKQDW-UHFFFAOYSA-N
Synonym 2-2-dimethylamino ethoxy benzylamine, 2-2-dimethylamino ethoxy phenyl methanamine, 2-2-aminomethyl phenoxy-n,n-dimethylethanamine, 2-2-aminomethyl phenoxy-n,n-dimethyl-ethanamine, 2-2-dimethylamino ethoxy benzenemethanamine, 2-2-aminomethyl phenoxy ethyl dimethylamine, n,n-dimethyl-2-2-aminomethyl phenoxy ethylamine, n-2-2-aminomethyl phenoxy ethyl-n,n-dimethylamine
PubChem CID 235943
IUPAC Name 2-[2-(aminomethyl)phenoxy]-N,N-dimethylethanamine
SMILES CN(C)CCOC1=CC=CC=C1CN

Specifications

CAS Min % 89.5
CAS Max % 100.0
Assay Percent Range 90%
Quantity 1 g
Formula Weight 194.28
Percent Purity 90%
Chemical Name or Material 2-[2-(Dimethylamino)ethoxy]benzylamine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.