missing translation for 'onlineSavingsMsg'
Learn More

2-[2-(Dimethylamino)ethoxy]benzoic acid, 95%, Thermo Scientific™

Product Code. 10293423
Change view
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10293423 250 mg Amber Glass Bottle 250mg
10469503 1 g Amber Glass Bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10293423 Supplier Thermo Scientific Maybridge Supplier No. CC51301CB
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 206261-66-7
Molecular Formula C11H15NO3
Molecular Weight (g/mol) 209.245
MDL Number MFCD07801180
InChI Key JZLYEZGHBQFRLU-UHFFFAOYSA-N
Synonym 2-2-dimethylamino ethoxy benzoic acid, 2-2-dimethylamino-ethoxy-benzoic acid, 2-2-dimethylaminoethoxy benzoic acid, 2-2-dimethylaminoethyloxy benzoic acid, 2-carboxy-beta-dimethylamino phenetole, 2-2-n,n-dimethylamino ethoxy benzoic acid, 2-2-n,n-dimethylamino ethyl oxy-benzoic acid
PubChem CID 6484659
IUPAC Name 2-[2-(dimethylamino)ethoxy]benzoic acid
SMILES CN(C)CCOC1=CC=CC=C1C(=O)O

Specifications

CAS Min % 94.5
CAS Max % 100.0
Assay Percent Range 95%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 209.24
Percent Purity 95%
Chemical Name or Material 2-[2-(Dimethylamino)ethoxy]benzoic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.