missing translation for 'onlineSavingsMsg'
Learn More

2-[2-(Dimethylamino)ethoxy]aniline, 97%, Thermo Scientific™

Product Code. 10340274
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10340274 1 g 1g
10401514 250 mg 250mg
10336954 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10340274 Supplier Thermo Scientific Maybridge Supplier No. CC51314DA
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 1202-00-2
Molecular Formula C10H16N2O
Molecular Weight (g/mol) 180.251
InChI Key OHDPNHWSANKRNF-UHFFFAOYSA-N
Synonym 2-2-dimethylamino ethoxy aniline, 2-2-dimethylamino-ethoxy-phenylamine, benzenamine,2-2-dimethylamino ethoxy, benzenamine, 2-2-dimethylamino ethoxy, 2-2-aminophenoxy ethyl dimethylamine, 2-2-dimethylaminoethyloxy aniline
PubChem CID 6484656
IUPAC Name 2-[2-(dimethylamino)ethoxy]aniline
SMILES CN(C)CCOC1=CC=CC=C1N

Specifications

Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 180.25
Percent Purity 97%
Chemical Name or Material 2-[2-(Dimethylamino)ethoxy]aniline
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.