missing translation for 'onlineSavingsMsg'
Learn More

2,2-Dimethyl-propanoic Acid (6aR)-5,6,6a,7-Tetrahydro-10-hydroxy-6-methyl-4H-dibenzo[de,g]quinolin-11-yl Ester, TRC

Product Code. 30379979
Change view
Click to view available options
Quantity:
5 mg
Unit Size:
5mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30379979 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30379979 Supplier Toronto Research Chemicals Supplier No. TRCD4780305MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 2,2-Dimethyl-propanoic Acid (6aR)-5,6,6a,7-Tetrahydro-10-hydroxy-6-methyl-4H-dibenzo[de,g]quinolin-11-yl Ester
CAS 1350191-90-0
Molecular Formula C22H25NO3
Quantity 5 mg
Synonym (R)-10-Hydroxy-6-methyl-5,6,6a,7-tetrahydro-4H-dibenzo[de,g]quinolin-11-yl Pivalate
InChI Formula InChI=1S/C22H25NO3/c1-22(2,3)21(25)26-20-17(24)9-8-14-12-16-18-13(10-11-23(16)4)6-5-7-15(18)19(14)20/h5-9,16,24H,10-12H2,1-4H3/t16-/m1/s1
SMILES CN1CCc2cccc3c2[C@H]1Cc4ccc(O)c(OC(=O)C(C)(C)C)c34
IUPAC Name [(6aR)-10-hydroxy-6-methyl-5,6,6a,7-tetrahydro-4H-dibenzo[de,g]quinoline-11-yl] 2,2-dimethylpropanoate
Molecular Weight (g/mol) 351.44
Formula Weight 351.1834
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.