missing translation for 'onlineSavingsMsg'
Learn More

2,2-Dimethyl-5-oxo-1,3-dioxolane-4-acetic Acid Phenylmethyl Ester, TRC

Product Code. 30387766
Change view
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30387766 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30387766 Supplier Toronto Research Chemicals Supplier No. TRCD47662550MG
 Restricted Item
Estimated Shipment: 13-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Especificaciones

Chemical Name or Material 2,2-Dimethyl-5-oxo-1,3-dioxolane-4-acetic Acid Phenylmethyl Ester
CAS 1049703-02-7
Molecular Formula C14H16O5
Quantity 50 mg
InChI Formula InChI=1S/C14H16O5/c1-14(2)18-11(13(16)19-14)8-12(15)17-9-10-6-4-3-5-7-10/h3-7,11H,8-9H2,1-2H3
SMILES CC1(C)OC(CC(=O)OCc2ccccc2)C(=O)O1
IUPAC Name benzyl 2-(2,2-dimethyl-5-oxo-1,3-dioxolan-4-yl)acetate
Molecular Weight (g/mol) 264.27
Formula Weight 264.0998
Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.