missing translation for 'onlineSavingsMsg'
Learn More

2,2-Dimethyl-4-pentenal, 97%, stabilized

Product Code. 10017402 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 mL
Packaging:
Glass bottle
Unit Size:
5mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10017402 5 mL Glass bottle 5mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10017402 Supplier Thermo Scientific Acros Supplier No. 295040050
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 5497-67-6 | C7H12O | 112.17 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

CAS 5497-67-6
CAS Min % 96.0
CAS Max % 100.0
Color Colorless to Yellow
Density 0.8250g/mL
Boiling Point 124.0°C to 125.0°C
Flash Point 18°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Molecular Formula C7H12O
Linear Formula H2C=CHCH2C(CH3)2CHO
Refractive Index 1.4200 to 1.4220
MDL Number MFCD00010699
Quantity 5 mL
Specific Gravity 0.825
Synonym 2,2-dimethyl-4-pentenal, 4-pentenal, 2,2-dimethyl, 2,2-dimethyl-pent-4-enal, 4-pentenal,2,2-dimethyl, zlchem 644, acmc-20aojn, pubchem22726, 2,2-dimethyl4-pentenal, 4-pentenal,2-dimethyl, 2,2-dimethy-4-pentenal
InChI Key DXSDIWHOOOBQTJ-UHFFFAOYSA-N
SMILES CC(C)(CC=C)C=O
IUPAC Name 2,2-dimethylpent-4-enal
Stabilizer 0.1% hydroquinone
Molecular Weight (g/mol) 112.17
PubChem CID 79636
Formula Weight 112.17
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material 2, 2-Dimethyl-4-pentenal
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 2,2-Dimethyl-4-pentenal, stabilized
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H225-Highly flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P240-Ground/bond container and receiving equipment.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.