Learn More
(2,2-Dimethyl-2,3-dihydro-1-benzofuran-7-yl)methylamine, ≥97%, Maybridge™
Brand: Maybridge CC27213CB
Code : 31
Additional Details : CAS Number : 868755-44-6 Weight : 0.00025kg Transport : UN number : 2735 Chem class : 8 Pack group : III
Packaging | Amber Glass Bottle |
---|---|
Quantity | 250mg |
Chemical Identifiers
868755-44-6 | |
177.247 | |
BQEPNISHAAOSFF-UHFFFAOYSA-N | |
18525713 | |
CC1(CC2=C(O1)C(=CC=C2)CN)C |
C11H15NO | |
MFCD08271885 | |
2,2-dimethyl-2,3-dihydro-1-benzofuran-7-yl methylamine, 2,2-dimethyl-2,3-dihydrobenzofuran-7-yl methanamine, 2,2-dimethyl-3h-1-benzofuran-7-yl methanamine, 1-2,2-dimethyl-3h-1-benzofuran-7-yl methanamine, 7-benzofuranmethanamine,2,3-dihydro-2,2-dimethyl, 2,2-dimethyl-2,3-dihydro-1-benzofuran-7-yl methanamine, 2,3-dihydro-2,2-dimethyl-1-benzofuran-7-yl methylamine, 2,2-dimethyl-2,3-dihydrobenzo 2,3-b furan-7-yl methylamine, 1-2,2-dimethyl-2,3-dihydro-1-benzofuran-7-yl methanamine, 2,2-dimethyl-2,3-dihydro-1-benzo b furan-7-yl methylamine | |
(2,2-dimethyl-3H-1-benzofuran-7-yl)methanamine |
Specifications
(2, 2-Dimethyl-2, 3-dihydro-1-benzofuran-7-yl)methylamine | |
250mg | |
96.5 | |
97% | |
MFCD08271885 | |
BQEPNISHAAOSFF-UHFFFAOYSA-N | |
(2,2-dimethyl-3H-1-benzofuran-7-yl)methanamine | |
18525713 | |
97% |
Amber Glass Bottle | |
868755-44-6 | |
100.0 | |
C11H15NO | |
2,2-dimethyl-2,3-dihydro-1-benzofuran-7-yl methylamine, 2,2-dimethyl-2,3-dihydrobenzofuran-7-yl methanamine, 2,2-dimethyl-3h-1-benzofuran-7-yl methanamine, 1-2,2-dimethyl-3h-1-benzofuran-7-yl methanamine, 7-benzofuranmethanamine,2,3-dihydro-2,2-dimethyl, 2,2-dimethyl-2,3-dihydro-1-benzofuran-7-yl methanamine, 2,3-dihydro-2,2-dimethyl-1-benzofuran-7-yl methylamine, 2,2-dimethyl-2,3-dihydrobenzo 2,3-b furan-7-yl methylamine, 1-2,2-dimethyl-2,3-dihydro-1-benzofuran-7-yl methanamine, 2,2-dimethyl-2,3-dihydro-1-benzo b furan-7-yl methylamine | |
CC1(CC2=C(O1)C(=CC=C2)CN)C | |
177.247 | |
177.25 |