missing translation for 'onlineSavingsMsg'
Learn More

2,2-Dimethyl-2,3-dihydro-1-benzofuran-7-yl isocyanate, 97%, Thermo Scientific™

Catalog Number p-3522432
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
87254-55-5
C11H11NO2
189.214
MFCD09025825
JTJCJMYVKXVWIJ-UHFFFAOYSA-N
2,2-dimethyl-2,3-dihydro-1-benzofuran-7-yl isocyanate, 7-isocyanato-2,2-dimethyl-2,3-dihydro-1-benzofuran, 2,2-dimethyl-2,3-dihydrobenzo b furan-7-isocyanate, benzofuran,2,3-dihydro-7-isocyanato-2,2-dimethyl
18525712
7-isocyanato-2,2-dimethyl-3H-1-benzofuran
CC1(CC2=C(O1)C(=CC=C2)N=C=O)C
This item is not returnable. View return policy
87254-55-5
C11H11NO2
189.214
MFCD09025825
JTJCJMYVKXVWIJ-UHFFFAOYSA-N
2,2-dimethyl-2,3-dihydro-1-benzofuran-7-yl isocyanate, 7-isocyanato-2,2-dimethyl-2,3-dihydro-1-benzofuran, 2,2-dimethyl-2,3-dihydrobenzo b furan-7-isocyanate, benzofuran,2,3-dihydro-7-isocyanato-2,2-dimethyl
18525712
7-isocyanato-2,2-dimethyl-3H-1-benzofuran
CC1(CC2=C(O1)C(=CC=C2)N=C=O)C
CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 189.21
Percent Purity 97%
Chemical Name or Material 2, 2-Dimethyl-2, 3-dihydro-1-benzofuran-7-yl isocyanate
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'