missing translation for 'onlineSavingsMsg'
Learn More

2,2-Dimethyl-2,3-dihydro-1-benzofuran-7-carbonyl chloride, 97%, Thermo Scientific™

Product Code. 10232592
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10232592 1 g Amber Glass Bottle 1g
10591862 250 mg Amber Glass Bottle 250mg
10183404 5 g Amber Glass Bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10232592 Supplier Thermo Scientific Maybridge Supplier No. CC27202DA
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 499785-51-2
Molecular Formula C11H11ClO2
Molecular Weight (g/mol) 210.657
MDL Number MFCD04115386
InChI Key LSSXEVHVRBTNAQ-UHFFFAOYSA-N
Synonym 2,2-dimethyl-2,3-dihydro-1-benzofuran-7-carbonyl chloride, 2,2-dimethyl-2,3-dihydrobenzofuran-7-carbonyl chloride, 2,2-dimethyl-2,3-dihydrobenzo b furan-7-carbonyl chloride, 2,2-dimethyl-2,3-dihydro-1-benzo b furan-7-carbonyl chloride
PubChem CID 2795470
IUPAC Name 2,2-dimethyl-3H-1-benzofuran-7-carbonyl chloride
SMILES CC1(CC2=CC=CC(=C2O1)C(=O)Cl)C

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 210.66
Percent Purity 97%
Chemical Name or Material 2, 2-Dimethyl-2, 3-dihydro-1-benzofuran-7-carbonyl chloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.