missing translation for 'onlineSavingsMsg'
Learn More

2,2-Dimethyl-2,3-dihydro-1-benzofuran-7-carbaldehyde, 97%, Thermo Scientific™

Product Code. 10242592
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10242592 1 g Amber Glass Bottle 1g
10631293 250 mg Amber Glass Bottle 250mg
10470934 5 g Amber Glass Bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10242592 Supplier Thermo Scientific Maybridge Supplier No. CC27204DA
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 38002-88-9
Molecular Formula C11H12O2
Molecular Weight (g/mol) 176.22
MDL Number MFCD04115387
InChI Key HGKYVFOYQUSRQN-UHFFFAOYSA-N
Synonym 2,2-dimethyl-2,3-dihydro-1-benzofuran-7-carbaldehyde, 2,2-dimethyl-2,3-dihydrobenzofuran-7-carbaldehyde, 7-benzofurancarboxaldehyde, 2,3-dihydro-2,2-dimethyl, 2,2-dimethyl-7-formyl-2,3-dihydrobenzofuran, 2,2-dimethyl-2,3-dihydro-benzofuran-7-carbaldehyde, 2,3-dihydro-2,2-dimethylbenzofuran-7-carbaldehyde, 2,2-dimethyl-2,3-dihydrobenzo b furan-7-carbaldehyde, 2,3-dihydro-2,2-dimethylbenzofuran-7-carboxaldehyde, 2,2-dimethyl-2,3-dihydro-1-benzo b furan-7-carbaldehyde
PubChem CID 2795471
SMILES CC1(C)CC2=CC=CC(C=O)=C2O1

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
IUPAC Name 2,2-dimethyl-3H-1-benzofuran-7-carbaldehyde
Formula Weight 176.21
Percent Purity 97%
Chemical Name or Material 2, 2-Dimethyl-2, 3-dihydro-1-benzofuran-7-carbaldehyde
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.