missing translation for 'onlineSavingsMsg'
Learn More

2,2'-Dimethyl-[1,1'-biphenyl]-4,4'-diamine, TRC

Product Code. 30387753
Change view
Click to view available options
Quantity:
250 mg
500 mg
2.5 g
Unit Size:
2.5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30387753 2.5 g 2.5g
30378442 500 mg -
30371394 250 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30387753 Supplier Toronto Research Chemicals Supplier No. TRCD4601132.5G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 84-67-3
Molecular Formula C14H16N2
Molecular Weight (g/mol) 212.29
Synonym 2,2'-Dimethylbenzidine, [1,1'-Biphenyl]-4,4'-diamine, 2,2'-dimethyl-, Benzidine, 2,2'-dimethyl- (7CI,8CI), 2,2'-Dimethyl[1,1'-biphenyl]-4,4'-diamine, 2,2'-Dimethyl-4,4'-biphenyldiamine, 2,2'-Dimethyl-4,4'-diaminobenzidine, 2,2'-Dimethyl-4,4'-diaminobiphenyl, 2,2'-Dimethyl-4,4'-diaminobiphenylene, 2,2'-Dimethylbenzidine, 2,2'-Tolidine, 4,4'-Diamino-2,2'-dimethylbiphenyl, 4-(4-Amino-2-methylphenyl)-3-methylaniline, DMBZ, NSC 1989, m-TB, m-TBHG, m-Tolidine
SMILES Cc1cc(N)ccc1c2ccc(N)cc2C

Specifications

Quantity 2.5 g
InChI Formula InChI=1S/C14H16N2/c1-9-7-11(15)3-5-13(9)14-6-4-12(16)8-10(14)2/h3-8H,15-16H2,1-2H3
Formula Weight 212.1313
Chemical Name or Material 2,2'-Dimethyl-[1,1'-biphenyl]-4,4'-diamine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.