missing translation for 'onlineSavingsMsg'
Learn More

2,2-Dimethoxypropane, 98+%

Product Code. 10410384 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
500 mL
1 L
2.5 L
10 L
Packaging:
Glass bottle
Metal Drum
Unit Size:
1 L
10 L
2.5 L
500mL
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10410384 2.5 L Glass bottle 2.5 L
10274330 500 mL Glass bottle 500mL
10214290 1 L Glass bottle 1 L
11994761 10 L Metal Drum 10 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 10410384 Supplier Thermo Scientific Acros Supplier No. 115630025
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 77-76-9 | C5H12O2 | 104.15 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Alkylation reagent
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 77-76-9
Molecular Formula C5H12O2
Molecular Weight (g/mol) 104.15
InChI Key HEWZVZIVELJPQZ-UHFFFAOYSA-N
Synonym acetone dimethyl acetal, propane, 2,2-dimethoxy, acetone dimethyl ketal, acetone, dimethyl acetal, acetone-dimethyl acetal, 2,2-dimethoxy propane, acetone dimethylacetal, 2,2-dimethyoxypropane, 2,2-dimethoxy-propane, dimethoxypropan
PubChem CID 6495
IUPAC Name 2,2-dimethoxypropane
SMILES CC(C)(OC)OC

Specifications

CAS Min % 98.0
CAS Max % 100.0
Melting Point -47.0°C
Color Colorless
Density 0.8400g/mL
Boiling Point 83.0°C
Flash Point −5°C
Infrared Spectrum Authentic
Assay Percent Range 98% min. (GC)
Packaging Glass bottle
Linear Formula (CH3)2C(OCH3)2
Refractive Index 1.3760 to 1.3780
Quantity 2.5 L
Beilstein 01,648
Fieser 01,268; 05,226; 13,199; 15,138
Specific Gravity 0.84
Solubility Information Solubility in water: 18 g/100g (25°C). Other solubilities: soluble in acetone, benzene, ccl4, ether,, n-heptane and methanol
Viscosity 0.452 mPa.s (25°C)
Formula Weight 104.15
Percent Purity 98+%
Physical Form Liquid
Chemical Name or Material 2,2-Dimethoxypropane
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.