missing translation for 'onlineSavingsMsg'
Learn More

2,2'-Dimethoxybiphenyl, 97%

Product Code. 11431027
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11431027 100 g 100g
11411027 5 g 5g
11421027 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11431027 Supplier Thermo Scientific Alfa Aesar Supplier No. B23411.22
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 4877-93-4 | C14H14O2 | 214.26 g/mol

2,2-Dimethoxy-1,1-biphenyl is used in the synthesis of biphenyl scaffolds in the preparation of biphenyl-tetrathiafulvalene derivatives.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,2-Dimethoxy-1,1-biphenyl is used in the synthesis of biphenyl scaffolds in the preparation of biphenyl-tetrathiafulvalene derivatives.

Solubility
Soluble in chloroform, methanol.

Notes
Store in a cool, dry conditions, in well sealed conditions. Store away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4877-93-4
Molecular Formula C14H14O2
Molecular Weight (g/mol) 214.26
MDL Number MFCD00067073
InChI Key VGMKUVCDINAAFC-UHFFFAOYSA-N
PubChem CID 611816
IUPAC Name 1-methoxy-2-(2-methoxyphenyl)benzene
SMILES COC1=CC=CC=C1C1=CC=CC=C1OC

Specifications

Melting Point 152°C to 156°C
Quantity 100 g
Synonym 2,2'-dimethoxybiphenyl, 2,2'-dimethoxy-1,1'-biphenyl, 1,1'-biphenyl, 2,2'-dimethoxy, 1-methoxy-2-2-methoxyphenyl benzene, 2,2'-dimethoxy-biphenyl, biphenyl, 2,2'-dimethoxy, 2,2/'-dimethoxybiphenyl, acmc-1ao09, 1,1'-biphenyl,2,2'-dimethoxy, 2,2'-dimethoxy-1,1'-biphenyl #
Solubility Information Soluble in chloroform,methanol.
Formula Weight 214.26
Percent Purity 97%
Chemical Name or Material 2,2'-Dimethoxybiphenyl

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.