missing translation for 'onlineSavingsMsg'
Learn More

2,2-Dimethoxy-N-methylethanamine, Mikromol™

Product Code. 30052508
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
This item is not returnable. View return policy

Product Code. 30052508

Brand: Mikromol MM0889.05

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

2,2-Dimethoxy-N-methylethanamine
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 2,2-Dimethoxy-N-methylethanamine
CAS 122-07-6
Analyte or Component Names 2,2-Dimethoxy-N-methylethanamine
Shipping Condition Room Temperature
Molecular Formula C5 H13 N O2
Certifications/Compliance ISO 9001
Synonym 2,2-Dimethoxy-N-methylethanamine, Methimazole Related Compound A, Thiamazole Impurity A
InChI Formula InChI=1S/C5H13NO2/c1-6-4-5(7-2)8-3/h5-6H,4H2,1-3H3
SMILES CNCC(OC)OC
IUPAC Name 2,2-dimethoxy-N-methylethanamine
Molecular Weight (g/mol) 119.16
Quantity 100 mg
Formula Weight 119.0946 g/mol
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.