missing translation for 'onlineSavingsMsg'
Learn More

2,2-Difluorosuccinic acid, 94%

Product Code. 11340204
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11340204 5 g 5g
11330204 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11340204 Supplier Thermo Scientific Alfa Aesar Supplier No. L00773.06
 Restricted Item
Estimated Shipment: 04-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 665-31-6 | C4H4F2O4 | 154.07 g/mol

2,2-Difluorosuccinic acid is used in the preparation of fluorofumaric acid. It is also used in enolate alkylation.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,2-Difluorosuccinic acid is used in the preparation of fluorofumaric acid. It is also used in enolate alkylation.

Solubility
Soluble in water.

Notes
Store in a cool place. Incompatible with strong oxidizing agents, strong acids, and strong bases.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 665-31-6
Molecular Formula C4H4F2O4
Molecular Weight (g/mol) 154.07
MDL Number MFCD00040970
InChI Key ZYLFHISLYSHWRH-UHFFFAOYSA-N
Synonym 2,2-difluorosuccinic acid, 2,2-difluorosuccinicacid, butanedioic acid, 2,2-difluoro, pubchem23428, 2,2-difluoro-butanedioic acid, butanedioic acid,2,2-difluoro, 2,2-difluoro succinic acid, 2,2-difluorobutane-1,4-dioic acid
PubChem CID 69581
IUPAC Name 2,2-difluorobutanedioic acid
SMILES OC(=O)CC(F)(F)C(O)=O

Specifications

Melting Point 139°C to 144°C
Flash Point >110°C (230°F)
Quantity 5 g
UN Number UN3261
Beilstein 1773355
Solubility Information Soluble in water.
Formula Weight 154.07
Percent Purity 94%
Chemical Name or Material 2,2-Difluorosuccinic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.