missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,2-Difluoropropionic acid, TRC
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
Specifications
Specifications
| Chemical Name or Material | 2,2-Difluoropropionic acid |
| CAS | 373-96-6 |
| Molecular Formula | C3H4O2F2 |
| Quantity | 100 mg |
| InChI Formula | InChI=1S/C3H4F2O2/c1-3(4,5)2(6)7/h1H3,(H,6,7) |
| SMILES | CC(F)(F)C(=O)O |
| IUPAC Name | 2,2-difluoropropanoic acid |
| Molecular Weight (g/mol) | 110.05 |
| Formula Weight | 110.0179 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction