missing translation for 'onlineSavingsMsg'
Learn More

2,2-Difluorocyclopropylamine hydrochloride, 97%

Product Code. 11442220
Click to view available options
Quantity:
100 mg
500 mg
Unit Size:
100mg
500mg
This item is not returnable. View return policy

Product Code. 11442220

Brand: Thermo Scientific Alfa Aesar H29051.MC

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 105614-25-3 | C3H6ClF2N | 129.535 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 105614-25-3
Molecular Formula C3H6ClF2N
Molecular Weight (g/mol) 129.535
MDL Number MFCD07777150
InChI Key WSSNWLHFBOKGGL-UHFFFAOYSA-N
Synonym 2,2-difluorocyclopropylamine hydrochloride, 2,2-difluorocyclopropan-1-amine hydrochloride, 2,2-difluorocycloprop-1-ylamine hydrochloride, 2,2-difluorocyclopropanamine hydrochloride, cyclopropanamine, 2,2-difluoro-, hydrochloride, 2,2-difluorocyclopropanamine, acmc-20aojv, 2,2-difluorocyclopropylamine, chloride, 2,2-difluorocycloprop-1-ylaminehydrochloride, 2,2-difluoro-1-cyclopropanamine hydrochloride
PubChem CID 20161313
IUPAC Name 2,2-difluorocyclopropan-1-amine;hydrochloride
SMILES C1C(C1(F)F)N.Cl

Specifications

Melting Point ∽145°C (decomposition)
Quantity 100 mg
Sensitivity Hygroscopic
Formula Weight 129.54
Percent Purity 97%
Chemical Name or Material 2,2-Difluorocyclopropylamine hydrochloride

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.