missing translation for 'onlineSavingsMsg'
Learn More

2,2-Difluorocyclopropanecarboxylic acid, 95%

Product Code. 11319969
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11319969 1 g 1g
11309969 250 mg 250mg
11329969 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11319969 Supplier Thermo Scientific Alfa Aesar Supplier No. H25752.03
 Restricted Item
Estimated Shipment: 03-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 107873-03-0 | C4H4F2O2 | 122.071 g/mol

2,2-Difluorocyclopropanecarboxylic acid derivative (i.e. 2,2-difluorocyclopropanecarbonyl chloride) is used in Friedel-Crafts reactions with various arenes to prepare aryl 2,2-difluorocyclopropyl ketones.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,2-Difluorocyclopropanecarboxylic acid derivative (i.e. 2,2-difluorocyclopropanecarbonyl chloride) is used in Friedel-Crafts reactions with various arenes to prepare aryl 2,2-difluorocyclopropyl ketones.

Solubility
Soluble in water.

Notes
Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 107873-03-0
Molecular Formula C4H4F2O2
Molecular Weight (g/mol) 122.071
MDL Number MFCD04115828
InChI Key KMLMOVWSQPHQME-UHFFFAOYSA-N
Synonym 2,2-difluorocyclopropanecarboxylic acid, 2,2-difluoro-cyclopropanecarboxylic acid, cyclopropanecarboxylic acid, 2,2-difluoro, acmc-20aht4, 2-carboxy-1,1-difluorocyclopropane, 2,2-difluoro cyclopropanecarboxylic acid, 2,2-difluoro-cyclopropanecarboxylic?acid, 2,2-difluorocyclopropane carboxylic acid, 2,2-difluorocyclopropane-carboxylic acid, 2,2-difluoro-cyclopropane-carboxylic acid
PubChem CID 2782823
IUPAC Name 2,2-difluorocyclopropane-1-carboxylic acid
SMILES C1C(C1(F)F)C(=O)O

Specifications

Melting Point 55°C to 60°C
Boiling Point 70°C (7 mmHg)
Quantity 1 g
UN Number UN3261
Solubility Information Soluble in water.
Formula Weight 122.07
Percent Purity 95%
Chemical Name or Material 2,2-Difluorocyclopropanecarboxylic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.