missing translation for 'onlineSavingsMsg'
Learn More

2,2-Difluoro-N-methoxy-N-methylacetamide, Thermo Scientific™

Product Code. 10403510
Change view
Click to view available options
Quantity:
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10403510 1 g Amber Glass Bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10403510 Supplier Thermo Scientific Maybridge Supplier No. MAY00153DA
 Restricted Item
Estimated Shipment: 07-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 142492-01-1
Packaging Amber Glass Bottle
Molecular Formula C4H7F2NO2
MDL Number MFCD06248152
Quantity 1 g
Synonym n-methyl-n-methoxydifluoroacetamide, acetamide, 2,2-difluoro-n-methoxy-n-methyl, acmc-20a3nv, n-methoxy-n-methyldifluoroacetamide, n-methoxy-n-methyl difluoroacetamide, n-difluoroacetyl-n-methoxymethylamine, acetamide,2,2-difluoro-n-methoxy-n-methyl, 2,2-bis fluoranyl-n-methoxy-n-methyl-ethanamide
InChI Key CUPRFYMJGQMIIC-UHFFFAOYSA-N
SMILES CON(C)C(=O)C(F)F
IUPAC Name 2,2-difluoro-N-methoxy-N-methylacetamide
Molecular Weight (g/mol) 139.10
PubChem CID 19023731
Formula Weight 139.1
Percent Purity 95%
Chemical Name or Material 2, 2-difluoro-N-methoxy-N-methylacetamide
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.