missing translation for 'onlineSavingsMsg'
Learn More

2,2-Difluoro-4-pentenoic acid, 97%

Product Code. 15434867
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15434867 5 g 5g
15444867 250 mg 250mg
15424867 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15434867 Supplier Thermo Scientific Alfa Aesar Supplier No. H32173.06
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 55039-89-9 | C5H6F2O2 | 136.098 g/mol

2,2-Difluoro-4-pentenoic acid is involved in the iodolactonization reaction to give the corresponding gamma- lactone, which is converted to prepare prepare 5-hydroxy-3,3-difluoropiperidine.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,2-Difluoro-4-pentenoic acid is involved in the iodolactonization reaction to give the corresponding gamma- lactone, which is converted to prepare prepare 5-hydroxy-3,3-difluoropiperidine.

Solubility
Immiscible with water.

Notes
Incompatible with oxidizing agents and bases.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 55039-89-9
Molecular Formula C5H6F2O2
Molecular Weight (g/mol) 136.098
MDL Number MFCD09800642
InChI Key LHOKYUDUAYXFGF-UHFFFAOYSA-N
Synonym 2,2-difluoropent-4-enoicacid, 2,2-difluoro-4-pentenoic acid, 4-pentenoic acid, 2,2-difluoro, 2,2-difluoro-pent-4-enoic acid, 10.14272/lhokyuduayxfgf-uhfffaoysa-n, doi:10.14272/lhokyuduayxfgf-uhfffaoysa-n
PubChem CID 11954656
IUPAC Name 2,2-difluoropent-4-enoic acid
SMILES C=CCC(C(=O)O)(F)F

Specifications

Boiling Point 56°C to 58°C (3 mmHg)
Quantity 5 g
UN Number UN3265
Solubility Information Immiscible with water.
Formula Weight 136.1
Percent Purity 97%
Chemical Name or Material 2,2-Difluoro-4-pentenoic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.