missing translation for 'onlineSavingsMsg'
Learn More

2,2-Difluoro-3-hydroxy-4-methyl-pentanoic acid, TRC

Product Code. 30382029
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30382029 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30382029 Supplier Toronto Research Chemicals Supplier No. TRCD4473651G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 2,2-Difluoro-3-hydroxy-4-methyl-pentanoic acid
CAS 1248233-69-3
Molecular Formula C6 H10 F2 O3
Quantity 1 g
InChI Formula InChI=1S/C6H10F2O3/c1-3(2)4(9)6(7,8)5(10)11/h3-4,9H,1-2H3,(H,10,11)
SMILES CC(C)C(O)C(F)(F)C(=O)O
IUPAC Name 2,2-difluoro-3-hydroxy-4-methylpentanoic acid
Molecular Weight (g/mol) 168.139
Formula Weight 168.06
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.