missing translation for 'onlineSavingsMsg'
Learn More

2,2-Difluoro-2-(fluorosulfonyl)acetic Acid, TRC

Product Code. 30372180
Change view
Click to view available options
Quantity:
10 g
25 g
Unit Size:
10g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30372180 25 g 25g
30380156 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30372180 Supplier Toronto Research Chemicals Supplier No. TRCD44611025G

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 2,2-Difluoro-2-(fluorosulfonyl)acetic Acid
CAS 1717-59-5
Molecular Formula C2 H F3 O4 S
Quantity 25 g
Synonym Difluoro(fluorosulfonyl)-acetic Acid, (Fluorosulfonyl)difluoroacetic Acid, 2-(Fluorosulfonyl)-2,2-difluoroacetic Acid, Difluoro(fluorosulfonyl)acetic Acid
InChI Formula InChI=1S/C2HF3O4S/c3-2(4,1(6)7)10(5,8)9/h(H,6,7)
SMILES OC(=O)C(F)(F)S(=O)(=O)F
IUPAC Name 2,2-difluoro-2-fluorosulfonylacetic acid
Molecular Weight (g/mol) 178.087
Formula Weight 177.955
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.