missing translation for 'onlineSavingsMsg'
Learn More

2’,2’-Difluoro-2’-deoxyuridine 3',5'-Dibenzoate, TRC

Product Code. 30384163
Change view
Click to view available options
Quantity:
25 mg
250 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30384163 25 mg
30376038 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30384163 Supplier Toronto Research Chemicals Supplier No. TRCD44574325MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 143157-27-1
Molecular Formula C23 H18 F2 N2 O7
Molecular Weight (g/mol) 472.395
Synonym 3,5-Dibenzoate-2,2-difluorouridine
IUPAC Name [(2R,5R)-3-benzoyloxy-5-(2,4-dioxopyrimidin-1-yl)-4,4-difluorooxolan-2-yl]methyl benzoate
SMILES FC1(F)C(OC(=O)c2ccccc2)[C@@H](COC(=O)c3ccccc3)O[C@H]1N4C=CC(=O)NC4=O

Specifications

Quantity 25 mg
InChI Formula InChI=1S/C23H18F2N2O7/c24-23(25)18(34-20(30)15-9-5-2-6-10-15)16(13-32-19(29)14-7-3-1-4-8-14)33-21(23)27-12-11-17(28)26-22(27)31/h1-12,16,18,21H,13H2,(H,26,28,31)/t16-,18?,21-/m1/s1
Formula Weight 472.108
Chemical Name or Material 2’,2’-Difluoro-2’-deoxyuridine 3',5'-Dibenzoate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.