missing translation for 'onlineSavingsMsg'
Learn More

2,2'-Dichlorobenzil, 98+%

Product Code. 11362546
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11362546 25 g 25g
11352546 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11362546 Supplier Thermo Scientific Alfa Aesar Supplier No. L06507.14
Spotlight Offers
Use promo code 28038
View all promotions
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 21854-95-5 | C14H8Cl2O2 | 279.116 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 21854-95-5
Molecular Formula C14H8Cl2O2
Molecular Weight (g/mol) 279.116
MDL Number MFCD00018263
InChI Key VOSNNSVWVJFJCR-UHFFFAOYSA-N
Synonym 2,2'-dichlorobenzil, 1,2-bis 2-chlorophenyl ethane-1,2-dione, ethanedione, bis 2-chlorophenyl, 2,2'-dichlorodibenzoyl, benzil, 2,2'-dichloro, 1,2-ethanedione, 1,2-bis 2-chlorophenyl, bis 2-chlorophenyl ethane-1,2-dione, benzil,2'-dichloro, 2,2/'-dichlorobenzil, acmc-20aok0
PubChem CID 89076
IUPAC Name 1,2-bis(2-chlorophenyl)ethane-1,2-dione
SMILES C1=CC=C(C(=C1)C(=O)C(=O)C2=CC=CC=C2Cl)Cl

Specifications

Melting Point 132°C to 136°C
Quantity 25 g
Beilstein 2120689
Formula Weight 279.12
Percent Purity ≥98%
Chemical Name or Material 2,2'-Dichlorobenzil

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.