missing translation for 'onlineSavingsMsg'
Learn More

2,2'-Dichlorobenzil, 98+%, Thermo Scientific Chemicals

Catalog Number 11352546
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy
21854-95-5
C14H8Cl2O2
279.116
MFCD00018263
VOSNNSVWVJFJCR-UHFFFAOYSA-N
2,2'-dichlorobenzil, 1,2-bis 2-chlorophenyl ethane-1,2-dione, ethanedione, bis 2-chlorophenyl, 2,2'-dichlorodibenzoyl, benzil, 2,2'-dichloro, 1,2-ethanedione, 1,2-bis 2-chlorophenyl, bis 2-chlorophenyl ethane-1,2-dione, benzil,2'-dichloro, 2,2/'-dichlorobenzil, acmc-20aok0
89076
1,2-bis(2-chlorophenyl)ethane-1,2-dione
C1=CC=C(C(=C1)C(=O)C(=O)C2=CC=CC=C2Cl)Cl
This item is not returnable. View return policy
21854-95-5
C14H8Cl2O2
279.116
MFCD00018263
VOSNNSVWVJFJCR-UHFFFAOYSA-N
2,2'-dichlorobenzil, 1,2-bis 2-chlorophenyl ethane-1,2-dione, ethanedione, bis 2-chlorophenyl, 2,2'-dichlorodibenzoyl, benzil, 2,2'-dichloro, 1,2-ethanedione, 1,2-bis 2-chlorophenyl, bis 2-chlorophenyl ethane-1,2-dione, benzil,2'-dichloro, 2,2/'-dichlorobenzil, acmc-20aok0
89076
1,2-bis(2-chlorophenyl)ethane-1,2-dione
C1=CC=C(C(=C1)C(=O)C(=O)C2=CC=CC=C2Cl)Cl

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 132°C to 136°C
Quantity 5 g
Beilstein 2120689
Formula Weight 279.12
Percent Purity ≥98%
Chemical Name or Material 2,2'-Dichlorobenzil

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'