missing translation for 'onlineSavingsMsg'
Learn More

2,2-Dichloroacetic Acid-1-13C, TRC

Product Code. 30387194
Change view
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30387194 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30387194 Supplier Toronto Research Chemicals Supplier No. TRCD43182810MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 2,2-Dichloroacetic Acid-1-13C
CAS 173470-70-7
Purity Grade Notes HPLC
Molecular Formula C13CH2Cl2O2
Quantity 10 mg
Synonym Dichloroacetic acid 13C (C1-13C), Acetic-1-13C acid, 2,2-dichloro- (9CI)
InChI Formula InChI=1S/C2H2Cl2O2/c3-1(4)2(5)6/h1H,(H,5,6)/i2+1
SMILES O[13C](C(Cl)Cl)=O
Molecular Weight (g/mol) 129.93
Percent Purity >95
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.