missing translation for 'onlineSavingsMsg'
Learn More

2,2-Dichloro-1,3-benzodioxole, 97%

Product Code. 11320016
Change view
Click to view available options
Quantity:
1 g
10 g
Unit Size:
10g
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11320016 1 g 1g
11310016 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11320016 Supplier Thermo Scientific Alfa Aesar Supplier No. H32056.03

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 2032-75-9 | C7H4Cl2O2 | 191.01 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2032-75-9
Molecular Formula C7H4Cl2O2
Molecular Weight (g/mol) 191.01
MDL Number MFCD00196818
InChI Key NMNKOUVBELKKTR-UHFFFAOYSA-N
Synonym 1,3-benzodioxole,2,2-dichloro, 2,2-dichlorobenzo d 1,3 dioxole, dcmdb, 1,2-dichloromethylenedioxy benzene
PubChem CID 11041568
IUPAC Name 2,2-dichloro-2H-1,3-benzodioxole
SMILES ClC1(Cl)OC2=CC=CC=C2O1

Specifications

Density 1.54
Boiling Point 215°C
Flash Point 85°C (185°F)
Quantity 1 g
UN Number UN3265
Sensitivity Moisture sensitive
Formula Weight 191.01
Percent Purity 97%
Chemical Name or Material 2,2-Dichloro-1,3-benzodioxole

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.