missing translation for 'onlineSavingsMsg'
Learn More

2,2'-Dibromobiphenyl, TRC

Product Code. 30376427
Change view
Click to view available options
Quantity:
1 g
5 g
10 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30376427 10 g
30373834 1 g
30378806 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30376427 Supplier Toronto Research Chemicals Supplier No. TRCD42555810G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13029-09-9
Molecular Formula C12 H8 Br2
Molecular Weight (g/mol) 312
Synonym 1,1'-Biphenyl, 2,2'-dibromo- (9CI, ACI), 2,2'-Dibromo-1,1'-biphenyl (ACI), Biphenyl, 2,2'-dibromo- (6CI, 7CI, 8CI), 1-Bromo-2-(2-bromophenyl)benzene, 2,2'-Dibromobiphenyl, 2,2'-Dibromodiphenyl, NSC 91566, o,o'-Dibromobiphenyl, PBB 4
IUPAC Name 1-bromo-2-(2-bromophenyl)benzene
SMILES Brc1ccccc1c2ccccc2Br

Specifications

Quantity 10 g
InChI Formula InChI=1S/C12H8Br2/c13-11-7-3-1-5-9(11)10-6-2-4-8-12(10)14/h1-8H
Formula Weight 309.8993
Chemical Name or Material 2,2'-Dibromobiphenyl
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.