missing translation for 'onlineSavingsMsg'
Learn More

2,2'-Diaminodiphenyl disulfide, 97%

Product Code. p-7037169
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
This item is not returnable. View return policy

Product Code. 15482158

Brand: Thermo Scientific Alfa Aesar H55807.03

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 1141-88-4 | C12H12N2S2 | 248.36 g/mol

2,2'-Diaminodiphenyl disulfide, is used as an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff. It is also used as a primary and secondary intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,2′-Diaminodiphenyl disulfide, is used as an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff. It is also used as a primary and secondary intermediate.

Solubility
Soluble in hot methanol- almost transparent.

Notes
Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1141-88-4
Molecular Formula C12H12N2S2
Molecular Weight (g/mol) 248.36
MDL Number MFCD00007703
InChI Key YYYOQURZQWIILK-UHFFFAOYSA-N
Synonym 2,2'-dithiodianiline, benzenamine, 2,2'-dithiobis, intramine, 2,2'-disulfanediyldianiline, 2-aminophenyl disulfide, 2,2'-diaminodiphenyldisulfide, 2,2'-diaminodiphenyl disulfide, 2-2-aminophenyl disulfanyl aniline, aniline, 2,2'-dithiodi, bis o-aminophenyl disulfide
PubChem CID 14358
IUPAC Name 2-[(2-aminophenyl)disulfanyl]aniline
SMILES NC1=CC=CC=C1SSC1=CC=CC=C1N

Specifications

Melting Point 91°C to 92°C
Quantity 1 g
Beilstein 914032
Solubility Information Soluble in hot methanol- almost transparent.
Formula Weight 248.36
Percent Purity 97%
Chemical Name or Material 2,2'-Diaminodiphenyl disulfide
compliance-icons compliance-icons compliance-icons
Product Identifier
  • 2,2'-Diaminodiphenyl disulfide
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • ACUTE AQUATIC Acute 1
  • Serious eye damage/eye irritation Category 1
Hazard Statement
  • H302-Harmful if swallowed.
  • H318-Causes serious eye damage.
  • H400-Very toxic to aquatic life.
  • H410-Very toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contains : 2,2'-dithiobisbenzenamine

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.