missing translation for 'onlineSavingsMsg'
Learn More

2,2-Di(tert-butylperoxy)butane, 50% solution in aromatic free mineral spirit

Product Code. 10499252 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
10 mL
500 mL
Packaging:
Plastic bottle
Unit Size:
10mL
500mL
This item is not returnable. View return policy

Product Code. 10499252

Brand: Thermo Scientific Acros 349835000

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 2167-23-9 | C12H26O4 | 234.336 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Radical initiators
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2167-23-9, 31807-55-3, 78-93-3
Molecular Formula C12H26O4
Molecular Weight (g/mol) 234.336
MDL Number MFCD00048243
InChI Key HQOVXPHOJANJBR-UHFFFAOYSA-N
Synonym 2,2-di tert-butylperoxy butane, 2,2-bis tert-butylperoxy butane, chaloxyd p 1293al, trigonox d, chaloxyd p 1200al, butane, 2,2-bis tert-butyldioxy, unii-v15k90hftj, v15k90hftj, 2,2-bis t-butylperoxy butane, di-tert-butyl sec-butylidene diperoxide
PubChem CID 221260
IUPAC Name 2,2-bis(tert-butylperoxy)butane
SMILES CCC(C)(OOC(C)(C)C)OOC(C)(C)C

Specifications

Color Undesignated
CAS Min % 49.0
CAS Max % 51.0
Density 0.8050g/mL
Solubility Information Solubility in water: immiscible. Other solubilities: miscible with saturated aliphatic and aromatic, hydrocarbons
Formula Weight 234.34
Percent Purity 49 to 51%
Packaging Plastic bottle
Specific Gravity 0.805
Viscosity 2.16 mPa.s (20°C)
Quantity 500 mL
Flash Point 43°C
Physical Form Solution
Chemical Name or Material 2, 2-Di(tert-butylperoxy)butane, 50% Solution in Aromatic Free Mineral Spirit
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 2,2-Di(tert-butylperoxy)butane, 50% solution in aromatic free mineral spirit
Signal Word
  • Danger
Hazard Category
  • Aspiration toxicity Category 1
  • Flammable liquid Category 3
  • Organic peroxide Type C & D
Hazard Statement
  • H304-May be fatal if swallowed and enters airways.
  • H242-Heating may cause a fire.
  • H226-Flammable liquid and vapour.
  • EUH066-Repeated exposure may cause skin dryness or cracking.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P220-Keep/Store away from clothing/combustible materials.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P331-Do NOT induce vomiting.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.