missing translation for 'onlineSavingsMsg'
Learn More

2-[2-(Di-N-propylamino)ethyl]-6-nitrophenyl Acetic Acid-N-oxide, TRC

Product Code. 30386679
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30386679 50 mg 50mg
30374944 100 mg -
30387472 10 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30386679 Supplier Toronto Research Chemicals Supplier No. TRCD48087050MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1072875-70-7
Molecular Formula C16H24N2O5
Molecular Weight (g/mol) 324.37
Synonym Proylidene Ropinirole Impurity, 2-(2-(2-(Dipropylamino)ethyl)-6-nitrophenyl)acetic Acid
SMILES CCC[N+]([O-])(CCC)CCc1cccc(c1CC(=O)O)[N+](=O)[O-]

Specifications

Quantity 50 mg
InChI Formula InChI=1S/C16H24N2O5/c1-3-9-18(23,10-4-2)11-8-13-6-5-7-15(17(21)22)14(13)12-16(19)20/h5-7H,3-4,8-12H2,1-2H3,(H,19,20)
Formula Weight 324.1685
Chemical Name or Material 2-[2-(Di-N-propylamino)ethyl]-6-nitrophenyl Acetic Acid-N-oxide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.