missing translation for 'onlineSavingsMsg'
Learn More

2,2-Di-n-propylacetyl chloride, 98%

Product Code. 11372746
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11372746 25 g 25g
11362746 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11372746 Supplier Thermo Scientific Alfa Aesar Supplier No. L08852.14
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 2936-08-5 | C8H15ClO | 162.66 g/mol

2,2-Di-n-propylacetyl chloride is used to produce 1,2-bis-(2-propyl-valerylamino)-ethan.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,2-Di-n-propylacetyl chloride is used to produce 1,2-bis-(2-propyl-valerylamino)-ethan.

Solubility
Reacts with water.

Notes
Moisture Sensitive. Incompatible materials are Oxidizing agents and bases. Store away from moisture and strong bases.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2936-08-5
Molecular Formula C8H15ClO
Molecular Weight (g/mol) 162.66
MDL Number MFCD00051446
InChI Key PITHYUDHKJKJNQ-UHFFFAOYSA-N
Synonym 2-propylvaleryl chloride, valproyl chloride, 2,2-di-n-propylacetyl chloride, dipropylacetyl chloride, 2-propylvaleroyl chloride, 2-n-propyl-n-valeroyl chloride, valproic acid chloride, di-n-propylacetyl chloride, acmc-2097cd, 2-n-propylvaleroyl chloride
PubChem CID 76240
IUPAC Name 2-propylpentanoyl chloride
SMILES CCCC(CCC)C(=O)Cl

Specifications

Boiling Point 68°C to 70°C (10 mmHg)
Quantity 25 g
UN Number UN3265
Beilstein 1098856
Formula Weight 162.66
Percent Purity 98%
Chemical Name or Material 2,2-Di-n-propylacetyl chloride

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.