missing translation for 'onlineSavingsMsg'
Learn More

2,2-Di-n-propylacetic acid, 98+%

Product Code. 11484120
Change view
Click to view available options
Quantity:
10 g
25 g
50 g
250 g
Unit Size:
10g
250g
25g
50g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11484120 25 g 25g
11459813 10 g 10g
11469813 50 g 50g
11479813 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 11484120 Supplier Thermo Scientific Alfa Aesar Supplier No. A12962.14
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 99-66-1 | C8H16O2 | 144.21 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 99-66-1
Molecular Formula C8H16O2
Molecular Weight (g/mol) 144.21
MDL Number MFCD00002672
InChI Key NIJJYAXOARWZEE-UHFFFAOYSA-N
Synonym valproic acid, dipropylacetic acid, depakene, depakine, 2-propylvaleric acid, ergenyl, di-n-propylacetic acid, valproate, mylproin, pentanoic acid, 2-propyl
PubChem CID 3121
ChEBI CHEBI:39867
IUPAC Name 2-propylpentanoic acid
SMILES CCCC(CCC)C(O)=O

Specifications

Boiling Point 219°C to 220°C
Flash Point 126°C (258°F)
Odor Weak, Characteristic
Quantity 25 g
UN Number UN3265
Beilstein 1750447
Merck Index 14,9913
Formula Weight 144.21
Percent Purity ≥98%
Chemical Name or Material 2,2-Di-n-propylacetic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.