missing translation for 'onlineSavingsMsg'
Learn More

2,2-Di-n-propylacetamide, 97%

Product Code. 11312746
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11312746 25 g 25g
11302746 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11312746 Supplier Thermo Scientific Alfa Aesar Supplier No. L08847.14
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 2430-27-5 | C8H17NO | 143.23 g/mol

It is employed as an antiepileptic drug. It is hydrolyzed quickly to VPA in vivo, but has intrinsic anticonvulsant activity. Also a reactant used in the synthesis of mutual prodrugs containing bio-cleavable and drug releasable disulfide linkers.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Solubility
Soluble in chloroform, dimethyl sulfoxide and methanol. Insoluble in water.

Notes
Store in cool. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2430-27-5
Molecular Formula C8H17NO
Molecular Weight (g/mol) 143.23
MDL Number MFCD00051534
InChI Key OMOMUFTZPTXCHP-UHFFFAOYSA-N
Synonym valpromide, depamide, dipropylacetamide, valpramide, 2-propylpentamide, 2-propylvaleramide, pentanamide, 2-propyl, 2,2-di-n-propylacetamide, 2-ethylvaleramide, propyl-2 valeramide
PubChem CID 71113
ChEBI CHEBI:74562
IUPAC Name 2-propylpentanamide
SMILES CCCC(CCC)C(N)=O

Specifications

Melting Point 123°C to 127°C
Odor Odorless
Quantity 25 g
Beilstein 1750444
Merck Index 14,9914
Solubility Information Soluble in chloroform,dimethyl sulfoxide and methanol. Insoluble in water.
Formula Weight 143.23
Percent Purity 97%
Chemical Name or Material 2,2-Di-n-propylacetamide

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.