missing translation for 'onlineSavingsMsg'
Learn More

2-(2-Chlorophenyl)-2,3-dihydrobenzo[d]oxazole-5-carboxylic Acid, TRC

Product Code. 30360175
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30360175 100 mg
30353427 10 mg
30354910 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30360175 Supplier Toronto Research Chemicals Supplier No. TRCC723015100MG
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1018498-30-0
Molecular Formula C14 H10 Cl N O3
Molecular Weight (g/mol) 275.687
Synonym 2-(2-Chlorophenyl)-1,3-Benzoxazole-5-Carboxylic Acid
IUPAC Name 2-(2-chlorophenyl)-2,3-dihydro-1,3-benzoxazole-5-carboxylic acid
SMILES OC(=O)c1ccc2OC(Nc2c1)c3ccccc3Cl

Specifications

Quantity 100 mg
InChI Formula InChI=1S/C14H10ClNO3/c15-10-4-2-1-3-9(10)13-16-11-7-8(14(17)18)5-6-12(11)19-13/h1-7,13,16H,(H,17,18)
Formula Weight 275.035
Chemical Name or Material 2-(2-Chlorophenyl)-2,3-dihydrobenzo[d]oxazole-5-carboxylic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.