missing translation for 'onlineSavingsMsg'
Learn More

2-(2-Chloroethoxy)ethanol, 99%

Product Code. 10656572 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
50 mL
250 mL
Packaging:
Glass bottle
Unit Size:
250mL
50mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10656572 250 mL Glass bottle 250mL
10194470 50 mL Glass bottle 50mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10656572 Supplier Thermo Scientific Acros Supplier No. 172262500
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 628-89-7 | C4H9ClO2 | 124.56 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 628-89-7
Molecular Formula C4H9ClO2
Molecular Weight (g/mol) 124.56
MDL Number MFCD00002870
InChI Key LECMBPWEOVZHKN-UHFFFAOYSA-N
Synonym 2-2-chloroethoxy ethanol, ethanol, 2-2-chloroethoxy, diglycol chlorohydrin, diglycol chlorhydrin, 2-chloroethyl 2-hydroxyethyl ether, monochloropolyoxyethylene, 2-2-chloroethoxy ethan-1-ol, unii-r2up818l4i, diethylene glycol monochlorohydrin, polyethylene glycol chloride 210
PubChem CID 12361
IUPAC Name 2-(2-chloroethoxy)ethanol
SMILES OCCOCCCl

Specifications

CAS Min % 98.5
CAS Max % 100.0
Density 1.1800g/mL
Boiling Point 180°C to 185°C
Flash Point 90°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Linear Formula ClCH2CH2OCH2CH2OH
Refractive Index 1.4519 to 1.4539
Quantity 250 mL
Beilstein 01, 467
Specific Gravity 1.18
Solubility Information Solubility in water: very soluble. Other solubilities: miscible with ethyl alcohol and diethyl ether
Formula Weight 124.57
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material 2-(2-Chloroethoxy)ethanol, 99%
Show More Show Less
compliance-icons
Product Identifier
  • 2-(2-Chloroethoxy)ethanol
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H319-Causes serious eye irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.