missing translation for 'onlineSavingsMsg'
Learn More

2-[2-Chloro-5-(trifluoromethyl)pyridin-4-ylamino]-N-methylbenzamide, TRC

Product Code. 30351538
Change view
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30351538 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30351538 Supplier Toronto Research Chemicals Supplier No. TRCC42198525MG
 Restricted Item
Estimated Shipment: 13-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 2-[2-Chloro-5-(trifluoromethyl)pyridin-4-ylamino]-N-methylbenzamide
CAS 1061358-71-1
Molecular Formula C14H11ClF3N3O
Quantity 25 mg
Synonym 2-[[2-Chloro-5-(trifluoromethyl)-4-pyridinyl]amino]-N-methylbenzamide
InChI Formula InChI=1S/C14H11ClF3N3O/c1-19-13(22)8-4-2-3-5-10(8)21-11-6-12(15)20-7-9(11)14(16,17)18/h2-7H,1H3,(H,19,22)(H,20,21)
SMILES CN=C(O)c1ccccc1Nc2cc(Cl)ncc2C(F)(F)F
IUPAC Name 2-[[2-chloro-5-(trifluoromethyl)pyridin-4-yl]amino]-N-methylbenzenecarboximidic acid
Molecular Weight (g/mol) 329.7
Formula Weight 329.0543
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.