missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-(2-Butoxyethoxy)ethanol, TRC
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
Specifications
Specifications
| Chemical Name or Material | Diethylene glycol-monobutyl ether |
| CAS | 112-34-5 |
| Molecular Formula | C8 H18 O3 |
| Quantity | 50 mg |
| Synonym | 2-(2-Butoxyethoxy)ethanol, 2-(2-n-Butoxyethoxy)ethanol, 3,6-Dioxa-1-decanol, BDG, BDG-NS, BDGE 20, Bikanol B 2, Butadigol, Butoxyethoxyethanol, Butycenol 20P, Butyl Carbitol, Butyl Diglysolv, Butyl Oxitol glycol ether, Butyl diglycol, Butyl digol, Butyl dioxitol, Butysenol 20, Butysenol 20P, C 4E2, Circuposit 3304, Circuposit 4125, Circuposit Hole Prep 3304, Circuposit Hole Prep 4125, DB, Diethylene glycol butyl ether, Diethylene glycol mono-n-butyl ether, Diethylene glycol monobutyl ether, Diethylene glycol n-butyl ether, Diglycol monobutyl ether, Dowanol DB, Ektasolve DB, Ethanol, 2,2'-oxybis-, monobutyl ether, Glycol Ether DB, Hisolve DB, K 50181, NBC, NBC (solvent), NSC 407762, O-Butyl diethylene glycol, Poly-Solv DB, Sinopol BDG-LS, n-Butyl carbitol |
| InChI Formula | InChI=1S/C8H18O3/c1-2-3-5-10-7-8-11-6-4-9/h9H,2-8H2,1H3 |
| SMILES | CCCCOCCOCCO |
| IUPAC Name | 2-(2-butoxyethoxy)ethanol |
| Molecular Weight (g/mol) | 162.23 |
| Formula Weight | 162.1256 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction